Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Pentadiene 95.0+%, TCI America™
SDP

Catalog No. P08051G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08051G 1 g
P08055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08051G Supplier TCI America Supplier No. P08051G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 591-95-7
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00039943
InChI Key LVMTVPFRTKXRPH-UHFFFAOYSA-N
Synonym Ethylallene
PubChem CID 11588
IUPAC Name penta-1,2-diene
SMILES CCC=C=C

Specifications

Color Colorless
Boiling Point 45°C
Quantity 1 g
UN Number 3295
Formula Weight 68.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Pentadiene
EINECSNumber (2)-0020
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.