Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Phenylethyl)piperazine, 98%

Catalog No. p-7028042
Click to view available options
Quantity:
10 g
50 g

CAS: 5321-49-3 | C12H18N2 | 190.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5321-49-3
Molecular Formula C12H18N2
Molecular Weight (g/mol) 190.29
MDL Number MFCD00040740
InChI Key LKUAPSRIYZLAAO-UHFFFAOYSA-N
Synonym 1-2-phenylethyl piperazine, 1-phenethylpiperazine, 1-phenethyl-piperazine, 1-2-phenylethyl-piperazine, 1-phenylethylpiperazine, 1-phenethyl piperazine, n-2-phenylethyl piperazine, piperazine, 1-2-phenylethyl, 2-phenylethyl piperazine, l-phenethylpiperazine
PubChem CID 79214
IUPAC Name 1-(2-phenylethyl)piperazine
SMILES C1CN(CCN1)CCC2=CC=CC=C2

Specifications

Density 1.01
Boiling Point 276°C
Flash Point >110°C (230°F)
Refractive Index 1.543
Quantity 10 g
UN Number UN2735
Beilstein 145893
Sensitivity Air Sensitive
Formula Weight 190.29
Percent Purity 98%
Chemical Name or Material 1-(2-Phenylethyl)piperazine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 226-186-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.