1,2-Propanediol, ACS, 99.5%, Alfa Aesar™

As a substitute for ethylene glycol and glycerol. Good solvent for resins. Dissolves many essential oils, but immiscible with fixed oils.

Manufacturer: Alfa Aesar 30948K2

Quantity 1L

 View more versions of this product

Catalog No. AA30948K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 57-55-6
    Molecular Formula C3H8O2
    Formula Weight 76.1
    MDL Number MFCD00064272
    Beilstein 1340498
    Merck Index 14,7855
    Synonym (+/-)-Propylene glycol; 1,2-Dihydroxypropane
    Chemical Name or Material 1,2-Propanediol
    Grade ACS Grade

    Specifications

    Formula Weight 76.1
    Quantity 1L
    Physical Form Liquid
    pH Neutral
    TSCA TSCA
    Molecular Formula C3H8O2
    Odor Nearly Odorless
    MDL Number MFCD00064272
    CAS 57-55-6
    Beilstein 1340498
    Merck Index 14,7855
    Refractive Index 1.4320
    Synonym (+/-)-Propylene glycol; 1,2-Dihydroxypropane
    Boiling Point 186° to 188°C
    Flash Point 107°C (225°F)
    Sensitivity Hygroscopic
    RTECS Number TY2000000
    Density 1.036
    Solubility Information Miscible with water, acetone, chloroform. Soluble in ether
    Melting Point −60°C
    Chemical Name or Material 1,2-Propanediol
    Assay 99.5%
    Grade ACS Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit