Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Propanedithiol 95.0+%, TCI America™
SDP

Catalog No. P082125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P082125ML 25 mL
P08215ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P082125ML Supplier TCI America Supplier No. P082125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 814-67-5
Molecular Formula C3H8S2
Molecular Weight (g/mol) 108.22
MDL Number MFCD00022073
InChI Key YGKHJWTVMIMEPQ-UHFFFAOYNA-N
Synonym 1,2-Dimercaptopropane, Propylenedithiol
PubChem CID 61217
IUPAC Name propane-1,2-dithiol
SMILES CC(S)CS

Specifications

Color Yellow
Quantity 25 mL
UN Number 1228
Formula Weight 108.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Propanedithiol
RTECSNumber TZ2585000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.