Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Pyrazinyl)piperazine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
34803-68-4
C8H12N4
164.21
MFCD00040766
HCGFLVDMFDHYJD-UHFFFAOYSA-N
2-piperazin-1-yl pyrazine,1-2-pyrazinyl-piperazine,1-piperazinylpyrazine,1-pyrazin-2-yl piperazine,1-2-pyrazinyl piperazine,pyrazin-2-yl-piperazine,3,4,5,6-tetrahydro-2h-1,2' bipyrazinyl,2-piperazinylpyrazine,2-piperazinopyrazine,1-piperazinyl pyrazine
2734639
2-piperazin-1-ylpyrazine
C1CN(CCN1)C2=NC=CN=C2
34803-68-4
C8H12N4
164.21
MFCD00040766
HCGFLVDMFDHYJD-UHFFFAOYSA-N
2-piperazin-1-yl pyrazine,1-2-pyrazinyl-piperazine,1-piperazinylpyrazine,1-pyrazin-2-yl piperazine,1-2-pyrazinyl piperazine,pyrazin-2-yl-piperazine,3,4,5,6-tetrahydro-2h-1,2' bipyrazinyl,2-piperazinylpyrazine,2-piperazinopyrazine,1-piperazinyl pyrazine
2734639
2-piperazin-1-ylpyrazine
C1CN(CCN1)C2=NC=CN=C2
Boiling Point 105°C to 108°C (0.5 mbar)
Color Yellow
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 164.21
Physical Form Crystalline Powder or Chunks
Percent Purity ≥97.5%
Chemical Name or Material 1-(2-Pyrazinyl)piperazine, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger