Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,2-Dioxathiolane 2-Oxide 98.0+%, TCI America™
SDP

Catalog No. D297725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D297725G 25 g
D29775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D297725G Supplier TCI America Supplier No. D297725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3741-38-6
Molecular Formula C2H4O3S
Molecular Weight (g/mol) 108.11
MDL Number MFCD00005354
InChI Key WDXYVJKNSMILOQ-UHFFFAOYSA-N
Synonym ethylene sulfite, glycol sulfite, 1,3,2-dioxathiolane, 2-oxide, glycolsulfite, 1,2-ethylene sulfite, cyclic ethylene sulfite, ethylene glycol cyclic sulfite, ethylene glycol, cyclic sulfite, ethylene sulphite, sulfurous acid, cyclic ester with ethylene glycol
PubChem CID 77342
IUPAC Name 1,3,2λ⁴-dioxathiolan-2-one
SMILES O=S1OCCO1

Specifications

Color Colorless
Boiling Point 172°C
Quantity 25 g
Formula Weight 108.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,2-Dioxathiolane 2-Oxide
EINECSNumber (5)-6552
RTECSNumber KW3900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.