Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Cyclohexanetricarbonitrile (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. C23275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23275G 5 g
C23271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23275G Supplier TCI America Supplier No. C23275G

Chemical Identifiers

CAS 183582-92-5
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.192
MDL Number MFCD08276327
InChI Key VMUOSHREZKXCIV-UHFFFAOYSA-N
Synonym 1,3,5-Tricyanocyclohexane
PubChem CID 11423741
IUPAC Name cyclohexane-1,3,5-tricarbonitrile
SMILES C1C(CC(CC1C#N)C#N)C#N

Specifications

Color White
Boiling Point 200°C
Quantity 5 g
UN Number 3439
Formula Weight 159.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Cyclohexanetricarbonitrile (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.