Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Tri(2-thienyl)benzene 98.0+%, TCI America™
SDP

Catalog No. T28661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28661G 1 g
T28665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28661G Supplier TCI America Supplier No. T28661G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15509-95-2
Molecular Formula C18H12S3
Molecular Weight (g/mol) 324.47
MDL Number MFCD02323391
InChI Key UBHPRZXDFVCNHZ-UHFFFAOYSA-N
PubChem CID 4121433
IUPAC Name 2-[3,5-bis(thiophen-2-yl)phenyl]thiophene
SMILES S1C=CC=C1C1=CC(=CC(=C1)C1=CC=CS1)C1=CC=CS1

Specifications

Melting Point 159°C
Color Yellow
Boiling Point 298°C
Quantity 1 g
Formula Weight 324.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tri(2-thienyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.