Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Triacryloylhexahydro-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. T1000100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1000100G 100 g
T100025G 25 g
T1000500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1000100G Supplier TCI America Supplier No. T1000100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 959-52-4
Molecular Formula C12H15N3O3
Molecular Weight (g/mol) 249.27
MDL Number MFCD00035738
InChI Key FYBFGAFWCBMEDG-UHFFFAOYSA-N
PubChem CID 70397
IUPAC Name 1-[3,5-di(prop-2-enoyl)-1,3,5-triazinan-1-yl]prop-2-en-1-one
SMILES C=CC(=O)N1CN(CN(C1)C(=O)C=C)C(=O)C=C

Specifications

Melting Point 156°C
Color White
Quantity 100 g
UN Number 2811
Formula Weight 249.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Triacryloylhexahydro-1,3,5-triazine
EINECSNumber (5)-1001
RTECSNumber XY9260000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.