Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Triaza-7-phosphaadamantane, 97+%, Thermo Scientific Chemicals

Catalog Number AAH6678506
Click to view available options
Quantity:
1 g
5 g
53597-69-6
C6H12N3P
157.157
MFCD00154905
FXXRPTKTLVHPAR-UHFFFAOYSA-N
1,3,5-triaza-7-phosphaadamantane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane, 1,3,5-triaza-7-phosphaadamantane,min, acmc-20ap5q, 1,5-triaza-9-phospha-adamantane, 1,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.13.7 decane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.1 3,7 decane
143061
C1N2CN3CN1CP(C2)C3
53597-69-6
C6H12N3P
157.157
MFCD00154905
FXXRPTKTLVHPAR-UHFFFAOYSA-N
1,3,5-triaza-7-phosphaadamantane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane, 1,3,5-triaza-7-phosphaadamantane,min, acmc-20ap5q, 1,5-triaza-9-phospha-adamantane, 1,5-triaza-7-phosphatricyclo 3.3.1.13,7 decane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.13.7 decane, 1,3,5-triaza-7-phosphatricyclo 3.3.1.1 3,7 decane
143061
C1N2CN3CN1CP(C2)C3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 244°C to 250°C
Color White
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 157.15
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 1,3,5-Triaza-7-phosphaadamantane
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit