Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Tribromobenzene, 98%

Catalog No. AAB2386022
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 626-39-1 | C6H3Br3 | 314.80 g/mol

1,3,5-Tribromobenzene is used as an intermediate for active pharmaceutical ingredient manufacturing. It acts as an intermediate for the preparation of 1,3,5-trimethoxytoluene. It forms molecular complexes with [60]-and [70] fullerenes. Further, it is employed as an internal standard during derivatization step in the determination of cyanide in human plasma and urine by gas chromatography-mass spectrometry. In addition to this, it reacts with the perfluoroalkenylzinc reagent in the presence of tetrakis(triphenylphosphine)palladium catalyst to yield a trifunctional monomer 1,3,5-tris(alfa, beta, beta-trifluorovinyl)benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Tribromobenzene is used as an intermediate for active pharmaceutical ingredient manufacturing. It acts as an intermediate for the preparation of 1,3,5-trimethoxytoluene. It forms molecular complexes with [60]-and [70] fullerenes. Further, it is employed as an internal standard during derivatization step in the determination of cyanide in human plasma and urine by gas chromatography-mass spectrometry. In addition to this, it reacts with the perfluoroalkenylzinc reagent in the presence of tetrakis(triphenylphosphine)palladium catalyst to yield a trifunctional monomer 1,3,5-tris(alfa, beta, beta-trifluorovinyl)benzene.

Solubility
Soluble in hot ethanol and acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-39-1
Molecular Formula C6H3Br3
Molecular Weight (g/mol) 314.80
MDL Number MFCD00000080
InChI Key YWDUZLFWHVQCHY-UHFFFAOYSA-N
Synonym benzene, 1,3,5-tribromo, 1,3,5-tribromo-benzene, unii-o3td0u1oaq, o3td0u1oaq, paragos 530416, 1,5-tribromobenzene, pubchem9590, 1,3,5-tribromobezene, benzene,3,5-tribromo, acmc-20a11o
PubChem CID 12279
IUPAC Name 1,3,5-tribromobenzene
SMILES BrC1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 121°C to 124°C
Boiling Point 271°C
Quantity 100 g
Beilstein 1858917
Solubility Information Soluble in hot ethanol and acetic acid. Insoluble in water.
Formula Weight 314.8
Percent Purity 98%
Chemical Name or Material 1,3,5-Tribromobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-947-3
RTECSNumber DC1957200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.