Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3,5-Tribromobenzene, 98%

Click to view available options
Quantity:
25g
100g
500g
626-39-1
C6H3Br3
314.802
MFCD00000080
YWDUZLFWHVQCHY-UHFFFAOYSA-N
benzene, 1,3,5-tribromo,1,3,5-tribromo-benzene,unii-o3td0u1oaq,o3td0u1oaq,paragos 530416,1,5-tribromobenzene,pubchem9590,1,3,5-tribromobezene,benzene,3,5-tribromo,acmc-20a11o
12279
1,3,5-tribromobenzene
C1=C(C=C(C=C1Br)Br)Br
626-39-1
C6H3Br3
314.802
MFCD00000080
YWDUZLFWHVQCHY-UHFFFAOYSA-N
benzene, 1,3,5-tribromo,1,3,5-tribromo-benzene,unii-o3td0u1oaq,o3td0u1oaq,paragos 530416,1,5-tribromobenzene,pubchem9590,1,3,5-tribromobezene,benzene,3,5-tribromo,acmc-20a11o
12279
1,3,5-tribromobenzene
C1=C(C=C(C=C1Br)Br)Br
Applications
1,3,5-Tribromobenzene is used as an intermediate for active pharmaceutical ingredient manufacturing. It acts as an intermediate for the preparation of 1,3,5-trimethoxytoluene. It forms molecular complexes with [60]-and [70] fullerenes. Further, it is employed as an internal standard during derivatization step in the determination of cyanide in human plasma and urine by gas chromatography-mass spectrometry. In addition to this, it reacts with the perfluoroalkenylzinc reagent in the presence of tetrakis(triphenylphosphine)palladium catalyst to yield a trifunctional monomer 1,3,5-tris(alfa, beta, beta-trifluorovinyl)benzene.

Solubility
Soluble in hot ethanol and acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Melting Point 121°C to 124°C
Boiling Point 271°C
Quantity 500g
Beilstein 1858917
Formula Weight 314.8
Percent Purity 98%
Chemical Name or Material 1,3,5-Tribromobenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DC1957200

TSCA : TSCA