Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,3,5-Tribromobenzene 98.0+%, TCI America™

Click to view available options
Quantity:
5 g
25 g
250 g
Specifications
Specifications
Melting Point | 122°C |
Boiling Point | 271°C |
Quantity | 250 g |
Formula Weight | 314.80 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 1,3,5-Tribromobenzene |
Safety and Handling
RTECSNumber | DC1957200 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction