Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Tribromobenzene 98.0+%, TCI America™
SDP

Catalog No. T034725G
Click to view available options
Quantity:
5 g
25 g
250 g

Chemical Identifiers

CAS 626-39-1
Molecular Formula C6H3Br3
Molecular Weight (g/mol) 314.80
MDL Number MFCD00000080
InChI Key YWDUZLFWHVQCHY-UHFFFAOYSA-N
Synonym benzene, 1,3,5-tribromo, 1,3,5-tribromo-benzene, unii-o3td0u1oaq, o3td0u1oaq, paragos 530416, 1,5-tribromobenzene, pubchem9590, 1,3,5-tribromobezene, benzene,3,5-tribromo, acmc-20a11o
PubChem CID 12279
IUPAC Name 1,3,5-tribromobenzene
SMILES BrC1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 122°C
Boiling Point 271°C
Quantity 25 g
Formula Weight 314.80
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tribromobenzene
RTECSNumber DC1957200
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.