Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Triethynylbenzene, 98%

Catalog No. p-7026425
Click to view available options
Quantity:
1 g
5 g

CAS: 7567-63-7 | C12H6 | 150.18 g/mol

1,3,5-Triethynylbenzene is used as the carbon precursor for graphene synthesis on rhodium. It is also used as a connecting unit for various transition metal building blocks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triethynylbenzene is used as the carbon precursor for graphene synthesis on rhodium. It is also used as a connecting unit for various transition metal building blocks.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7567-63-7
Molecular Formula C12H6
Molecular Weight (g/mol) 150.18
MDL Number MFCD00068621
InChI Key ZDRMMTYSQSIGRY-UHFFFAOYSA-N
PubChem CID 139048
IUPAC Name 1,3,5-triethynylbenzene
SMILES C#CC1=CC(=CC(=C1)C#C)C#C

Specifications

Melting Point 105°C to 107°C
Density 1.05
Quantity 1 g
UN Number UN1325
Synonym benzene, 1,3,5-triethynyl, benzene,1,3,5-triethynyl, 1,3,5-phenylene-triacetylene, acmc-209ozh, 1,3,5-triethynyl-benzene, 1,3,5-triethynylbenzene, 1,3,5-triethynylbenzene 1g, benzene, 1,3,5-triethynyl-8ci, 9ci
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 1,3,5-Triethynylbenzene
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.