Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Trifluorobenzene, 98+%

Catalog No. AAA1598414
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 372-38-3 | C6H3F3 | 132.085 g/mol

1,3,5-Trifluorobenzene was used in the synthesis of (-)-epicatechin and its 3-O-gallate. It is also used as an intermediate in organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trifluorobenzene was used in the synthesis of (-)-epicatechin and its 3-O-gallate. It is also used as an intermediate in organic chemical synthesis.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-38-3
Molecular Formula C6H3F3
Molecular Weight (g/mol) 132.085
MDL Number MFCD00000333
InChI Key JXUKFFRPLNTYIV-UHFFFAOYSA-N
Synonym benzene, 1,3,5-trifluoro, sym-trifluorobenzene, unii-bn94c411f8, pubchem1069, 1,5-trifluorobenzene, acmc-209iru, 2,4,6-trifluorophenyl, benzene,3,5-trifluoro, 1,3,5-tri-fluorobenzene, 1,3,5-trifluoro-benzene
PubChem CID 9745
IUPAC Name 1,3,5-trifluorobenzene
SMILES C1=C(C=C(C=C1F)F)F

Specifications

Melting Point -7°C to -4°C
Density 1.279
Boiling Point 75°C to 76°C
Flash Point −7°C (19°F)
Refractive Index 1.415
Quantity 25 g
UN Number UN2387
Beilstein 1906457
Solubility Information Insoluble in water.
Formula Weight 132.09
Percent Purity ≥98%
Chemical Name or Material 1,3,5-Trifluorobenzene
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROBENZENE
EINECSNumber 206-751-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.