Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth(III) oxyfluoride, 97%, pure, Thermo Scientific™

Catalog No. AC453821000
Click to view available options
Quantity:
25 g
100 g

Chemical Identifiers

CAS 13520-72-4
Molecular Formula BiFO
Molecular Weight (g/mol) 243.98

Specifications

Melting Point 500.0°C
Quantity 100 g
Assay Percent Range 96% min. (ex Bi)
Linear Formula FBi=O
Solubility Information Solubility in water: insoluble.
Formula Weight 243.98
Percent Purity 97%
Grade Pure
Chemical Name or Material Bismuth(III) oxyfluoride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep only in
EINECSNumber 236-861-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.