Learn More
1,3,5-Triisopropylbenzene, 95%
Description
1,3,5-Triisopropylbenzene is used as a fuel and fuel additive. It is also used in lubricants and lubricant additives. Further, it is used in the preparation of 2,4,6-triisopropyl-benzenesulfonyl chloride. It is also employed as a swelling agent during the synthesis of mesoporous silica with two-dimensional hexagonal pores. In addition to this, it is used as a micelle expander in the synthesis of magnetic mesoporous silica nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3,5-Triisopropylbenzene is used as a fuel and fuel additive. It is also used in lubricants and lubricant additives. Further, it is used in the preparation of 2,4,6-triisopropyl-benzenesulfonyl chloride. It is also employed as a swelling agent during the synthesis of mesoporous silica with two-dimensional hexagonal pores. In addition to this, it is used as a micelle expander in the synthesis of magnetic mesoporous silica nanoparticles.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | -7°C |
| Density | 0.854 |
| Boiling Point | 235°C to 237°C |
| Flash Point | 86°C (186°F) |
| Refractive Index | 1.488 |
| Quantity | 50 g |
| Beilstein | 1862750 |
| Solubility Information | Immiscible with water. |
| Formula Weight | 204.36 |
| Percent Purity | 95% |
| Show More |
Safety and Handling
| Hazard Category | H227-H311-H315-H319 |
| Hazard Statement | GHS H Statement H227 Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P361-P363-P370+P378q-P501c |
| EINECSNumber | 211-941-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.