Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Triisopropylbenzene 95.0+%, TCI America™
SDP

Catalog No. T0458100ML
Click to view available options
Quantity:
25 mL
100 mL
500 mL

Chemical Identifiers

CAS 717-74-8
Molecular Formula C15H24
Molecular Weight (g/mol) 204.357
MDL Number MFCD00008890
InChI Key VUMCUSHVMYIRMB-UHFFFAOYSA-N
Synonym 1,3,5-triisopropylbenzene, 2,4,6-triisopropylbenzene, benzene, 1,3,5-tris 1-methylethyl, 1,3,5-tris propan-2-yl benzene, unii-fr9y346wpb, benzene, 1,3,5-triisopropyl, fr9y346wpb, benzene, tris 1-methylethyl, pubchem13720, 1,5-triisopropylbenzene
PubChem CID 12860
IUPAC Name 1,3,5-tri(propan-2-yl)benzene
SMILES CC(C)C1=CC(=CC(=C1)C(C)C)C(C)C

Specifications

Color Colorless
Boiling Point 235°C
Quantity 100 mL
Formula Weight 204.36
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Triisopropylbenzene
EINECSNumber (3)-0022&(3)-3427
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.