Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Trimethyl-1H-pyrazol-4-amine, 97%, Thermo Scientific™

Catalog No. cd11242ea
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CD11242EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CD11242EA Supplier Thermo Scientific Chemicals Supplier No. CD11242EA
Only null left
Add to Cart

Chemical Identifiers

CAS 28466-21-9
Molecular Formula C6H11N3
Molecular Weight (g/mol) 125.175
MDL Number MFCD00052883
InChI Key SSDGMKHZMNTWLS-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazol-4-amine, 4-amino-1,3,5-trimethylpyrazole, trimethyl-1h-pyrazol-4-amine, pyrazole, 4-amino-1,3,5-trimethyl, 4-amino-1,3,5-trimethyl-1h-pyrazole, trimethylpyrazol-4-amine, 1,3,5-trimethyl-1h-pyrazol-4-ylamine, 1h-pyrazol-4-amine, 1,3,5-trimethyl, 1,3,5-trimethyl-1h-pyrazole-4-amine, 1,3,5-trimethylpyrazole-4-ylamine
PubChem CID 161603
IUPAC Name 1,3,5-trimethylpyrazol-4-amine
SMILES CC1=C(C(=NN1C)C)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 125.17
Percent Purity 97%
Chemical Name or Material 1, 3, 5-Trimethyl-1H-pyrazol-4-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.