Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene, 97%
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Quantity | 250 mg |
| UN Number | UN3261 |
| Beilstein | 2369523 |
| Synonym | 1,3,5-tris chloromethyl-2,4,6-trimethylbenzene, 1,3,5-tris-chloromethyl-2,4,6-trimethyl-benzene, benzene, 1,3,5-tris chloromethyl-2,4,6-trimethyl, ksc224a3n, tris-chloromethyl-mesitylene, 2,4,6-tris chloromethyl mesitylene, tris chloromethyl-2,4,6-trimethylbenzene, 1,3,5-trimethyl-2,4,6-tris chloromethyl benzene, benzene,3,5-tris chloromethyl-2,4,6-trimethyl, 1,3,5-tris chloromethyl-2,4,6-trimethyl-benzene |
| SMILES | CC1=C(C(=C(C(=C1CCl)C)CCl)C)CCl |
| PubChem CID | 291188 |
| Formula Weight | 265.6 |
| Percent Purity | 97% |
| Physical Form | Powder |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction