Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene, 97%

Click to view available options
Quantity:
250mg
1g
3863-11-4
C6H5F2N
129.11
MFCD00229016
AXNUZKSSQHTNPZ-UHFFFAOYSA-N
3,4-difluorobenzenamine,benzenamine, 3,4-difluoro,3,4-difluoro-phenylamine,3,4-difluorophenyl amine,3,4-difluoro aniline,3,4-difluorophenylamine,3,4-difluoranilin,3,4-difloroaniline,3,4-difluroaniline,3.4-difluoroaniline
77469
3,4-difluoroaniline
C1=CC(=C(C=C1N)F)F
3863-11-4
C6H5F2N
129.11
MFCD00229016
AXNUZKSSQHTNPZ-UHFFFAOYSA-N
3,4-difluorobenzenamine,benzenamine, 3,4-difluoro,3,4-difluoro-phenylamine,3,4-difluorophenyl amine,3,4-difluoro aniline,3,4-difluorophenylamine,3,4-difluoranilin,3,4-difloroaniline,3,4-difluroaniline,3.4-difluoroaniline
77469
3,4-difluoroaniline
C1=CC(=C(C=C1N)F)F
Quantity 1g
Color White
UN Number UN3261
Beilstein 2369523
Formula Weight 265.6
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 1,3,5-Tris(chloromethyl)-2,4,6-trimethylbenzene

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.