Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Adamantanedicarboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. A135825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A135825G 25 g
A13585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A135825G Supplier TCI America Supplier No. A135825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39269-10-8
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00167790
InChI Key PAVQGHWQOQZQEH-UHFFFAOYSA-L
Synonym 1,3-adamantanedicarboxylic acid, 1,3-dicarboxyadamantane, 1s,3s,5r,7r-adamantane-1,3-dicarboxylic acid, 1,3-adamantane dicarboxylic acid, tricyclo 3.3.1.1 sup 3,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.13,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1,3-dicarboxylic acid, 1r,3s,5s,7s-adamantane-1,3-dicarboxylic acid, 1,3-adamantanedicarboxylicacid
PubChem CID 64339
IUPAC Name adamantane-1,3-dicarboxylate
SMILES [O-]C(=O)C12CC3CC(C1)CC(C3)(C2)C([O-])=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 224.26
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Adamantanedicarboxylic Acid
RTECSNumber YD1994000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.