Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzenediboronic acid, 97%, Thermo Scientific™

Catalog No. p-7027853
Click to view available options
Quantity:
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,3-benzenediboronic acid is used to prepare 1,3-didydrxybenzene by oxidation reaction using alkaline hydrogen peroxide.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4612-28-6
Molecular Formula C6H8B2O4
Molecular Weight (g/mol) 165.746
MDL Number MFCD03092915
InChI Key UXPAASVRXBULRG-UHFFFAOYSA-N
Synonym 1,3-benzenediboronic acid, 1,3-phenylenediboronic acid, 1,3-benzendiboronic acid, 1,3-benzenediboronicacid, m-phenylenediboronic acid, 3-dihydroxyboranyl phenyl boronic acid, 3-dihydroxyboranyl phenylboronic acid, m-benzenediboronic acid, pubchem17415, 1,3-benzendibororic acid
PubChem CID 230477
IUPAC Name (3-boronophenyl)boronic acid
SMILES B(C1=CC(=CC=C1)B(O)O)(O)O

Specifications

Melting Point >300°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 165.75
Percent Purity 97%
Chemical Name or Material 1,3-Benzenediboronic acid
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.