Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzenedisulfonyl chloride, 97%

Catalog No. AAB2404414
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 585-47-7 | C6H4Cl2O4S2 | 275.11 g/mol

1,3-Benzenedisulfonyl chloride is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Benzenedisulfonyl chloride is used as a pharmaceutical intermediates.

Solubility
Soluble in DMSO and methanol. Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-47-7
Molecular Formula C6H4Cl2O4S2
Molecular Weight (g/mol) 275.11
MDL Number MFCD00041879
InChI Key ALIQZUMMPOYCIS-UHFFFAOYSA-N
Synonym 1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a
PubChem CID 96240
SMILES ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O

Specifications

Melting Point 58°C to 60°C
Quantity 25 g
UN Number UN3261
Beilstein 1118508
Sensitivity Moisture sensitive
Solubility Information Soluble in DMSO and methanol. Reacts with water.
IUPAC Name benzene-1,3-disulfonyl chloride
Formula Weight 275.13
Percent Purity 98%
Chemical Name or Material 1,3-Benzenedisulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 209-556-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.