Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzenedithiol, 95%

Catalog No. ac456100010
Click to view available options
Quantity:
1 g
5 g

CAS: 626-04-0 | C6H6S2 | 142.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-04-0
Molecular Formula C6H6S2
Molecular Weight (g/mol) 142.23
MDL Number MFCD00004842
InChI Key ZWOASCVFHSYHOB-UHFFFAOYSA-N
Synonym 1,3-benzenedithiol, 1,3-dimercaptobenzene, dithioresorcin, dithioresorcinol, 1,3-benzedithiol, acmc-20ap5g, ksc357g0j
PubChem CID 522062
IUPAC Name benzene-1,3-dithiol
SMILES SC1=CC(S)=CC=C1

Specifications

Melting Point 24.0°C
Density 1.2400g/mL
Boiling Point 128°C to 129°C (21.0 hPa)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula C6H4(SH)2
Quantity 1 g
Specific Gravity 1.24
Formula Weight 142.24
Percent Purity 95%
Chemical Name or Material 1,3-Benzenedithiol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc
EINECSNumber 210-925-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.