Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Benzothiazole-6-sulfonyl chloride, ≥95%, Thermo Scientific™

Catalog No. CC05903DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 181124-40-3
Molecular Formula C7H4ClNO2S2
Molecular Weight (g/mol) 233.684
MDL Number MFCD03086100
InChI Key XQOLJTWXFUSVOR-UHFFFAOYSA-N
Synonym benzo d thiazole-6-sulfonyl chloride, 1,3-benzothiazole-6-sulfonylchloride, 6-benzothiazolesulfonyl chloride, 1,3-benzothiazole-6-sulphonyl chloride, 6-chlorosulfonylbenzothiazole, buttpark 121\04-40, 1,3-benzothiazole, benzothiazol-6-ylchlorosulfone, 6-benzothiazolesulfonylchloride, benzothiazole-6-sulphonyl chloride
PubChem CID 2795170
IUPAC Name 1,3-benzothiazole-6-sulfonyl chloride
SMILES C1=CC2=C(C=C1S(=O)(=O)Cl)SC=N2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 233.7
Percent Purity 95%
Chemical Name or Material 1, 3-Benzothiazole-6-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.