Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(2-propynyloxy)benzene 97.0+%, TCI America™
SDP

Catalog No. B45211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45211G 1 g
B4521200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45211G Supplier TCI America Supplier No. B45211G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26627-36-1
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00224312
InChI Key HSRDPYWXTZGWFQ-UHFFFAOYSA-N
Synonym 1,3-Bis(propargyloxy)benzene
PubChem CID 11030523
IUPAC Name 1,3-bis(prop-2-ynoxy)benzene
SMILES C#CCOC1=CC(=CC=C1)OCC#C

Specifications

Melting Point 40°C
Color White-Yellow
Quantity 1 g
Formula Weight 186.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(2-propynyloxy)benzene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.