Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Bromo-2-fluorophenyl)ethanol, 98%, Thermo Scientific™

Catalog No. AAH6473606
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6473606 5 g
AAH64736MD 250 mg
AAH6473603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6473606 Supplier Thermo Scientific Chemicals Supplier No. H6473606

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1221715-80-5
Molecular Formula C8H8BrFO
Molecular Weight (g/mol) 219.05
MDL Number MFCD16140247
InChI Key QFVPIDFXSBXICP-UHFFFAOYNA-N
Synonym 1-3-bromo-2-fluorophenyl ethanol, 1-3-bromo-2-fluoro-phenyl ethanol, 1-3-bromo-2-fluorophenyl ethan-1-ol, 3-bromo-2-fluoro-alpha-methylbenzenemethanol
PubChem CID 53484284
IUPAC Name 1-(3-bromo-2-fluorophenyl)ethanol
SMILES CC(O)C1=C(F)C(Br)=CC=C1

Specifications

Quantity 5 g
Formula Weight 219.05
Percent Purity 98%
Chemical Name or Material 1-(3-Bromo-2-fluorophenyl)ethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.