Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Butadiene diepoxide, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019500
Click to view available options
Quantity:
250 mg

Specifications

CAS 1464-53-5
Vapor Pressure 25 mmHg (56°C)
Density 1.113 g/mL (at 25°C)
Flash Point 46°C
Physical Form Neat
Linear Formula C4H6O2
MDL Number MFCD00005120
Melting Point 2°C to 4°C
Boiling Point 56°C to 58°C (25 mmHg)
Synonym 1,2,3,4-Diepoxybutane
Type Analytical Standard
Quantity 250 mg
Formula Weight 86.09
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.