Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Cycloheptanedione, 97%, Thermo Scientific™

Catalog No. AC440450010
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1194-18-9
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.15
InChI Key DBOVMTXPZWVYAQ-UHFFFAOYSA-N
Synonym 1,3-cycloheptanedione, 1 3-cycloheptanedione 97, acmc-20aijy, 1,3-cycloheptandione, pubchem20261, cycloheptan-1,3-dione, cyclohep tane-1,3-dione, cycloheptane-1,3-quinone, cycloheptane-?1,?3-?dione, ksc173k8n
PubChem CID 4072367
IUPAC Name cycloheptane-1,3-dione
SMILES C1CCC(=O)CC(=O)C1

Specifications

Density 1.1000g/mL
Boiling Point 254.0°C
Flash Point >100°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Refractive Index 1.483
Quantity 1 g
Specific Gravity 1.1
Formula Weight 126.15
Percent Purity 97%
Chemical Name or Material 1,3-Cycloheptanedione
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.