Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Cyclohexanebis(methylamine), 99%, mixture of cis and trans, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
2579-20-6
C8H18N2
142.24
MFCD00001522
QLBRROYTTDFLDX-UHFFFAOYSA-N
1,3-cyclohexanedimethanamine,1,3-cyclohexanebis methylamine,1,3-bis aminomethyl cyclohexane,cyclohexane-1,3-diyldimethanamine,1,3-di aminomethyl cyclohexane,kodak silver halide solvent hs-103,cyclohexane, 1,3-bis aminomethyl,3-aminomethyl cyclohexyl methanamine,3-aminomethyl cyclohexyl methylamine
17406
[3-(aminomethyl)cyclohexyl]methanamine
C1CC(CC(C1)CN)CN
2579-20-6
C8H18N2
142.24
MFCD00001522
QLBRROYTTDFLDX-UHFFFAOYSA-N
1,3-cyclohexanedimethanamine,1,3-cyclohexanebis methylamine,1,3-bis aminomethyl cyclohexane,cyclohexane-1,3-diyldimethanamine,1,3-di aminomethyl cyclohexane,kodak silver halide solvent hs-103,cyclohexane, 1,3-bis aminomethyl,3-aminomethyl cyclohexyl methanamine,3-aminomethyl cyclohexyl methylamine
17406
[3-(aminomethyl)cyclohexyl]methanamine
C1CC(CC(C1)CN)CN
Specific Gravity 0.94
Flash Point 106°C
Boiling Point 220°C
Melting Point -70°C
Density 0.9400g/mL
Refractive Index 1.4920 to 1.494
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H10(CH2NH2)2
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in ethanol,ethyl ether
Formula Weight 142.24
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 1, 3-Cyclohexanebis(methylamine), 99%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 219-941-5

RTECSNumber : GU7000000

TSCA : TSCA