Alfa Aesar™ 1,3-Cyclohexanedione, 97+%, may cont. up to 1% NaCl

Manufacturer: Alfa Aesar™ A1110922

Quantity 100g

 View more versions of this product

Catalog No. AAA1110922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 504-02-9
    Molecular Formula C6H8O2
    Formula Weight 112.13
    MDL Number MFCD00001585
    Beilstein 385888
    Merck Index 14,3178
    Synonym Dihydroresorcinol
    Chemical Name or Material 1,3-Cyclohexanedione

    Specifications

    Formula Weight 112.13
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Recommended Storage 2°C to 8°C
    Molecular Formula C6H8O2
    MDL Number MFCD00001585
    CAS 504-02-9
    Beilstein 385888
    Merck Index 14,3178
    Melting Point 101° to 105°C
    Quantity 100g
    TSCA TSCA
    Synonym Dihydroresorcinol
    Concentration May contain up to 1% NaCl
    Chemical Name or Material 1,3-Cyclohexanedione
    Assay ≥97%

    Description

    Applications
    It is used as a nitisinone intermediate.

    Solubility
    Soluble in water (155 g/L) at 20°C, alcohol, chloroform, acetone, boiling benzene, ether (slightly), carbon disulfide (slightly), and petr ether (slightly).

    Notes
    Air & Light Sensitive. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.

    GHS H Statement
    H301
    Toxic if swallowed.

    2°C to 8°C

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit