Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Di-o-tolylguanidine, 99%

Catalog No. p-7028017
Click to view available options
Quantity:
100 g
500 g

CAS: 97-39-2 | C15H17N3 | 239.322 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-39-2
Molecular Formula C15H17N3
Molecular Weight (g/mol) 239.322
MDL Number MFCD00008513
InChI Key OPNUROKCUBTKLF-UHFFFAOYSA-N
Synonym 1,3-di-o-tolylguanidine, di-o-tolylguanidine, dotg, vulkacite dotg, ditolylguanidine, 1,3-ditolylguanidine, vulkacit dotg, eveite dotg, vulkacit dotg/c, nocceler dt
PubChem CID 7333
IUPAC Name 1,2-bis(2-methylphenyl)guanidine
SMILES CC1=CC=CC=C1NC(=NC2=CC=CC=C2C)N

Specifications

Melting Point 176°C to 178°C
Quantity 100 g
UN Number UN2811
Formula Weight 239.32
Percent Purity 99%
Chemical Name or Material 1,3-Di-o-tolylguanidine
Hazard Category H301
Hazard Statement GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 202-577-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.