Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Di-tert-butylimidazol-2-ylidene 98.0+%, TCI America™
SDP

Catalog No. D34721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34721G 1 g
D34725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34721G Supplier TCI America Supplier No. D34721G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 157197-53-0
Molecular Formula C11H20N2
Molecular Weight (g/mol) 180.30
MDL Number MFCD03788921
InChI Key FENRCIKTFREPGS-UHFFFAOYSA-N
Synonym ItBu
PubChem CID 10961644
IUPAC Name 1,3-di-tert-butyl-1H-imidazol-3-ium-2-ide
SMILES CC(C)(C)N1C=C[N+](=[C-]1)C(C)(C)C

Specifications

Quantity 1 g
Formula Weight 180.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Di-tert-butylimidazol-2-ylidene
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.