Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diacetylbenzene, 97%, Thermo Scientific Chemicals

Catalog No. p-7026082
Click to view available options
Quantity:
5 g

1,3-Diacetylbenzene was used in the preparation of polyhydroxylated analogs. It is an important raw material and intermediates in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diacetylbenzene was used in the preparation of polyhydroxylated analogs. It is an important raw material and intermediates in organic synthesis.

Solubility
Soluble in ethanol, benzene, chloroform.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6781-42-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00008740
InChI Key VCHOFVSNWYPAEF-UHFFFAOYSA-N
Synonym 1,3-diacetylbenzene, 1,1'-1,3-phenylene diethanone, ethanone, 1,1'-1,3-phenylene bis, m-diacetylbenzene, benzene-1,3-bis acetyl, m-acetylacetophenone, m-acetyl acetophenone, 1-3-acetylphenyl ethanone, 1-3-acetylphenyl ethan-1-one, m-diacetyl benzene
PubChem CID 23229
SMILES CC(=O)C1=CC(=CC=C1)C(C)=O

Specifications

Melting Point 35°C to 37°C
Boiling Point 150°C to 155°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2042511
Solubility Information Soluble in ethanol,benzene,chloroform.
IUPAC Name 1-(3-acetylphenyl)ethanone
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 1,3-Diacetylbenzene
EINECSNumber 229-842-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.