Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diaminopropane, 98%

Catalog No. AAA119320F
Click to view available options
Quantity:
100 mL
500 mL
2500 mL

CAS: 109-76-2 | C3H10N2 | 74.127 g/mol

1,3-Diaminopropane is commonly used in the preparation of cholinesterase inhibitors and thrombosis inhibitors. It is also used as a polymer cross linkage agent, polyurethane extender and spray coatings. It is involved in the preparation of agrochemicals and pharmaceutical intermediates. Further, it acts as a building block in the synthesis of heterocyclic compounds, which finds application in textile finishing. In addition to this, it is used as a ligand and form coordination complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diaminopropane is commonly used in the preparation of cholinesterase inhibitors and thrombosis inhibitors. It is also used as a polymer cross linkage agent, polyurethane extender and spray coatings. It is involved in the preparation of agrochemicals and pharmaceutical intermediates. Further, it acts as a building block in the synthesis of heterocyclic compounds, which finds application in textile finishing. In addition to this, it is used as a ligand and form coordination complexes.

Solubility
Miscible with water.

Notes
Hygroscopic. Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-76-2
Molecular Formula C3H10N2
Molecular Weight (g/mol) 74.127
MDL Number MFCD00008228
InChI Key XFNJVJPLKCPIBV-UHFFFAOYSA-N
Synonym 1,3-diaminopropane, 1,3-propanediamine, trimethylenediamine, 1,3-propylenediamine, propandiamine, alpha,omega-propanediamine, unii-cb3isl56kg, ccris 4054, 1,3-propane diamine, cb3isl56kg
PubChem CID 428
ChEBI CHEBI:15725
IUPAC Name propane-1,3-diamine
SMILES C(CN)CN

Specifications

Melting Point -12°C
Density 0.888
pH >12
Boiling Point 134°C to 136°C
Flash Point 48°C (118°F)
Odor Amine-like
Refractive Index 1.4573
Quantity 2500 mL
UN Number UN2734
Beilstein 605277
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Formula Weight 74.13
Percent Purity 98%
Chemical Name or Material 1,3-Diaminopropane
Show More Show Less
Hazard Category H226-H302-H310-H314-H317-H334-H335
Hazard Statement GHS H Statement
H310-H314-H318-H226-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: I
EINECSNumber 203-702-7
RTECSNumber TX6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.