Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diaminopropane Dihydrochloride 97.0+%, TCI America™
SDP

Catalog No. T0613100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0613100G 100 g
T061325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0613100G Supplier TCI America Supplier No. T0613100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10517-44-9
Molecular Formula C3H12Cl2N2
Molecular Weight (g/mol) 147.04
MDL Number MFCD00012525
InChI Key HYOCSVGEQMCOGE-UHFFFAOYSA-N
Synonym Trimethylenediamine Dihydrochloride, 1,3-Propanediamine Dihydrochloride
PubChem CID 165887
IUPAC Name dihydrogen propane-1,3-diamine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCCN

Specifications

Melting Point 253°C
Color White
Quantity 100 g
Formula Weight 147.04
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diaminopropane Dihydrochloride
EINECSNumber (2)-0149
RTECSNumber TX7392500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.