Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Dibromo-2-propanol

Catalog No. 3724045G Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-240-45G 5 g
37-240-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-240-45G Supplier Sigma Aldrich Supplier No. 3724045G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,3-dibromo-2-propanol is a dihalogenated alcohol. it is reported as bifunctional crosslinking reagent.1,3-dibromo-2-propanol may be used in the following studies:
  • To prepare the thioether ligand, used in the synthesis of palladium-phosphorus/sulfur nanoparticles
  • Preparation of 2-(alkoxy)propenyl bromide
  • Synthesis of 1,3-propanediamine derivatives connected to carbohydrates (sugar-pendant diamines)
  • Chemical crosslinking of nine single substitution cysteine mutants of staphylococcal nuclease
  • Preparation of 1,3-dinitrooxy-2-propanol, via reaction with agno3 in mecn at 80c

Specifications

CAS 96-21-9
Density 2.136 g/mL (at 25°C (literature))
Boiling Point 82°C to 83°C (7 mmHg)
Molecular Formula C3H6Br2O
Linear Formula BrCH2CH(OH)CH2Br
Refractive Index n20/D 1.552 (literature)
MDL Number MFCD00000216
Quantity 5 g
Molecular Weight (g/mol) 217.89
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.