Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibromobenzene, 97+%

Catalog No. p-7020682
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 108-36-1 | C6H4Br2 | 235.91 g/mol

1,3-Dibromobenzene is used in Suzuki coupling reaction for the preparation of [n]metacyclophanes. It is used to prepare 5-(3-bromo-phenyl)-2,4-di-tert-butoxy-pyrimidine using Pd(0) (PPh3)4 as a reagent. Further, it undergoes solid-supported potassium fluoride-aluminum oxide palladium-catalyzed polyarylation reaction with phenyl boronic acid to get conjugated polyaryls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dibromobenzene is used in Suzuki coupling reaction for the preparation of [n]metacyclophanes. It is used to prepare 5-(3-bromo-phenyl)-2,4-di-tert-butoxy-pyrimidine using Pd(0) (PPh3)4 as a reagent. Further, it undergoes solid-supported potassium fluoride-aluminum oxide palladium-catalyzed polyarylation reaction with phenyl boronic acid to get conjugated polyaryls.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-36-1
Molecular Formula C6H4Br2
Molecular Weight (g/mol) 235.91
MDL Number MFCD00000078
InChI Key JSRLURSZEMLAFO-UHFFFAOYSA-N
Synonym m-dibromobenzene, benzene, 1,3-dibromo, benzene, m-dibromo, unii-74ef6kh8tc, 1,3-dibromo-benzene, 74ef6kh8tc, 1,3-dibromo benzene, 3,5-dibromobenzene, 1,3-dibrormobenzene, 1, 3-dibromobenzene
PubChem CID 7927
ChEBI CHEBI:37151
IUPAC Name 1,3-dibromobenzene
SMILES BrC1=CC(Br)=CC=C1

Specifications

Melting Point °C to 7°C
Density 1.955
Boiling Point 219°C to 220°C
Flash Point 93°C (199°F)
Assay Percent Range 97%
Refractive Index 1.608
Quantity 25 g
UN Number UN3082
Beilstein 1904538
Solubility Information Immiscible with water.
Formula Weight 235.91
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 1, 3-Dibromobenzene
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H227
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 203-574-2
RTECSNumber CZ1790000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.