Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxyphenylacetone, 98+%

Catalog No. AC189802500
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle

CAS: 122-84-9 | C10H12O2 | 164.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-84-9
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00008773
InChI Key WFWKNGZODAOLEO-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetone, p-acetonylanisole, anisketone, 1-4-methoxyphenyl propan-2-one, 2-propanone, 1-4-methoxyphenyl, anisyl methyl ketone, p-methoxyphenylacetone, 4-methoxybenzyl methyl ketone, 1-p-methoxyphenyl-2-propanone, anisic ketone
PubChem CID 31231
IUPAC Name 1-(4-methoxyphenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(C=C1)OC

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Green-Yellow to Yellow
Density 1.0670g/mL
Boiling Point 266°C to 268°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5200 to 1.53
Linear Formula CH3OC6H4CH2COCH3
Quantity 250 g
Beilstein 08, IV, 460
Specific Gravity 1.067
Formula Weight 164.2
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenylacetone, 98+%
Show More Show Less
EINECSNumber 204-578-7
RTECSNumber UC2990000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.