Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloro-2-butene, Mixture of cis and trans Isomers 96%, Thermo Scientific™

Catalog No. AC406780050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926-57-8
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 124.99
MDL Number MFCD00000861
InChI Key WLIADPFXSACYLS-DUXPYHPUSA-N
Synonym 1,3-dichlorobutylene, unii-pwg1e5mrlj, 1,3-dichloro-2-butene, trans-1,3-dichlorobutene-2, 1,3-dichlorobut-2-ene, pwg1e5mrlj, 2-butene, 1,3-dichloro-, e, 2-butene, 1,3-dichloro, e-1,3-dichloro-2-butene, 1,3-dichlorobutene-2
PubChem CID 643911
IUPAC Name (2E)-1,3-dichlorobut-2-ene
SMILES C\C(Cl)=C/CCl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.16g/mL
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Refractive Index 1.4690 to 1.4730 (20°C, 589nm)
Linear Formula ClCH2CHCHClCH3
Quantity 5 g
Fieser 01,214; 02,111; 10,134
Formula Weight 125
Percent Purity ≥95% (sum of cis and trans isomers)
Physical Form Liquid
Chemical Name or Material 1, 3-Dichloro-2-butene isomers, 96%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS P Statement
Toxic if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.