Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloro-2-propanol, 98+%

Catalog No. p-7022987
Click to view available options
Quantity:
250 g
1000 g

CAS: 96-23-1 | C3H6Cl2O | 128.98 g/mol

1,3-Dichloro-2-propanol biotransformation to epichlorohydrin by the whole cells of recombinant Escherichia coli via resin-based in situ product removal has been investigated. It is used in the synthesis of glycerol, in the production of plastics and textiles and in the synthesis of pharmaceuticals. Also used as a solvent and as a cement for celluloid. It is also present in some foods, e.g. soy sauce, soup spices and instant soups, from hydrolization of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dichloro-2-propanol biotransformation to epichlorohydrin by the whole cells of recombinant Escherichia coli via resin-based in situ product removal has been investigated. It is used in the synthesis of glycerol, in the production of plastics and textiles and in the synthesis of pharmaceuticals. Also used as a solvent and as a cement for celluloid. It is also present in some foods, e.g. soy sauce, soup spices and instant soups, from hydrolization of proteins.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-23-1
Molecular Formula C3H6Cl2O
Molecular Weight (g/mol) 128.98
MDL Number MFCD00000951
InChI Key DEWLEGDTCGBNGU-UHFFFAOYSA-N
Synonym 1,3-dichloro-2-propanol, dichlorohydrin, 1,3-dichloroisopropanol, 1,3-dichlorohydrin, glycerol 1,3-dichlorohydrin, 2-propanol, 1,3-dichloro, enodrin, 1,3-dichloro-2-hydroxypropane, 1,3-dichloroisopropyl alcohol, sym-glycerol dichlorohydrin
PubChem CID 7289
ChEBI CHEBI:18917
IUPAC Name 1,3-dichloropropan-2-ol
SMILES OC(CCl)CCl

Specifications

Melting Point -4°C
Density 1.366
Boiling Point 174°C to 176°C
Flash Point 85°C (185°F)
Refractive Index 1.483
Linear Formula ClCH2CH(OH)CH2Cl
Quantity 250 g
UN Number UN2750
Beilstein 1732063
Merck Index 14,3074
Solubility Information Fully miscible in water.
Formula Weight 128.99
Percent Purity ≥98%
Chemical Name or Material 1,3-Dichloro-2-propanol
Show More Show Less
missing translation for 'healthHazard1' H227-H301-H312-H350
missing translation for 'healthHazard2' GHS H Statement
H301-H311-H350-H227
Toxic if swallowed.
Toxic in contact with skin.
May cause cancer.
Combustible liquid.
missing translation for 'healthHazard3' P201-P202-P210-P264b-P270-P281-P301+P310-P302+P352-P308+P313-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,3-DICHLOROPROPANOL-2
EINECSNumber 202-491-9
RTECSNumber UB1400000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.