Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloro-5-(chloromethyl)benzene, 97%, Thermo Scientific™

Catalog No. spb00444da
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 3290-06-0
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
InChI Key ZFLRKAMKGYNFPH-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzyl chloride, 1,3-dichloro-5-chloromethyl benzene, benzene, 1,3-dichloro-5-chloromethyl, 3,5-dichlorobenzylchloride, unii-c076i19eql, 5-chloromethyl-1,3-dichlorobenzene, pubchem16695, alpha,3,5-trichlorotoluene, 3,4-dichloro benzyl chloride, ksc495g8l
PubChem CID 137880
IUPAC Name 1,3-dichloro-5-(chloromethyl)benzene
SMILES C1=C(C=C(C=C1Cl)Cl)CCl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3CH2Cl
Quantity 1 g
Formula Weight 195.48
Percent Purity 97%
Chemical Name or Material 1, 3-Dichloro-5-(chloromethyl)benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.