Learn More
1,3-Dichloroacetone, 96%
Description
1,3-Dichloroacetone is used in the synthesis of citric acid. It is used as a solvent and as an intermediate in organic synthesis. Further, it serves as an acceptor substrate in the cross-aldol reaction with donor substrates such as acetone, cyclopentanone and cyclohexanone. It is also used in the preparation of an active pharmaceutical ingredient, famotidine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3-Dichloroacetone is used in the synthesis of citric acid. It is used as a solvent and as an intermediate in organic synthesis. Further, it serves as an acceptor substrate in the cross-aldol reaction with donor substrates such as acetone, cyclopentanone and cyclohexanone. It is also used in the preparation of an active pharmaceutical ingredient, famotidine.
Solubility
Soluble in water, ethanol and diethyl ether.
Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents, strong bases and reducing agents.
Specifications
Specifications
| Melting Point | 40°C to 44°C |
| Density | 1.383 |
| pH | 4.2 |
| Boiling Point | 172°C to 173°C |
| Flash Point | 95°C (203°F) |
| Odor | Pungent |
| Refractive Index | 1.44 |
| Quantity | 100 g |
| UN Number | UN2649 |
| Beilstein | 605456 |
| Show More |
Safety and Handling
| Hazard Category | H300+H310+H330-H314-H335-H340 |
| Hazard Statement | GHS H Statement H300-H310-H330-H314-H318-H341 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Suspected of causing genetic defects. |
| Precautionary Statement | P201-P202-P260-P262-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,3-DICHLOROACETONE |
| EINECSNumber | 208-585-6 |
| RTECSNumber | UC1430000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.