Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloroacetone, typically 99%, Thermo Scientific Chemicals

Catalog No. p-7049760
Click to view available options
Quantity:
100 g

1,3-Dichloroacetone was used as acceptor substrate in the cross-aldol reaction with donor substrates such as acetone, cyclopentanone and cyclohexanon

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-07-6
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.96
MDL Number MFCD00000937
InChI Key SUNMBRGCANLOEG-UHFFFAOYSA-N
Synonym 1,3-dichloroacetone, 1,3-dichloro-2-propanone, 1,3-dichloropropanone, s-dichloroacetone, sym-dichloroacetone, bis chloromethyl ketone, 2-propanone, 1,3-dichloro, acetone, 1,3-dichloro, chloromethyl ketone
PubChem CID 10793
IUPAC Name 1,3-dichloropropan-2-one
SMILES ClCC(=O)CCl

Specifications

Melting Point 43°C to 45°C
Density 1.3834 g/mL at 46°C
Boiling Point 173°C
Flash Point 89°C (193°F)
Odor Odorless
Refractive Index 1.44
Quantity 100 g
UN Number UN2649
Beilstein 605456
Merck Index 14,3052
Solubility Information Soluble in alcohol,diethyl ether,water.
Formula Weight 126.97
Percent Purity 99% (typically)
Physical Form Crystalline
Chemical Name or Material 1,3-Dichloroacetone
Show More Show Less
Hazard Category H300+H310+H330-H314-H335-H340
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318-H341
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P260-P262-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,3-DICHLOROACETONE
EINECSNumber 208-585-6
RTECSNumber UC1430000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.