Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichlorobenzene, 98%

Catalog No. p-103931
Click to view available options
Quantity:
5 mL
25 mL
250 mL
500 mL
1 L
Packaging:
Glass bottle

CAS: 541-73-1 | C6H4Cl2 | 147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-73-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 147
MDL Number MFCD00000573
InChI Key ZPQOPVIELGIULI-UHFFFAOYSA-N
Synonym m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene
PubChem CID 10943
ChEBI CHEBI:36693
IUPAC Name 1,3-dichlorobenzene
SMILES C1=CC(=CC(=C1)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -24°C
Color Colorless to Yellow
Density 1.2800g/mL
Boiling Point 172°C to 173°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5450 to 1.547
Linear Formula C6H4Cl2
Quantity 25 mL
Beilstein 05, 202
Specific Gravity 1.28
Merck Index 15, 3064
Solubility Information Solubility in water: 0.0123g/100mL water (25°C). Other solubilities: soluble in organic solvents such as alcohol,,benzene,ether
Viscosity 1.045 mPa.s (23°C)
Formula Weight 147
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 3-Dichlorobenzene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-792-1
RTECSNumber CZ4499000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.