Learn More
1,3-Dichlorotetramethyldisiloxane, 96%
Description
1,3-Dichlorotetramethyldisiloxane is used to produce 1,1,3,3-tetramethyl-disiloxane-1,3-diol. It is also used as Chemical Additives. Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3-Dichlorotetramethyldisiloxane is used to produce 1,1,3,3-tetramethyl-disiloxane-1,3-diol. It is also used as Chemical Additives. Pharmaceutical intermediates.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from strong bases and water. Protect from water and humidity. Incompatible with acids, bases and oxidizing agents. Avoid excessive heat and light.
Specifications
Specifications
| Boiling Point | 136°C to 138°C |
| Melting Point | -37°C |
| Quantity | 25 g |
| UN Number | UN2985 |
| Beilstein | 1741218 |
| Flash Point | 15°C (59°F) |
| Solubility Information | Reacts with water. |
| Formula Weight | 203.22 |
| Percent Purity | 96% |
| Refractive Index | 1.408 |
| Show More |
Safety and Handling
| Hazard Category | H225-H314 |
| Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S. |
| EINECSNumber | 219-278-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.