Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diethylbenzene, 97+%

Catalog No. aa43369ab
Click to view available options
Quantity:
2 mL
10 mL

CAS: 141-93-5 | C10H14 | 134.222 g/mol

1,3-Diethylbenzene is used as an intermediate and solvent. It is also used in the preparation of divinyl benzene. Further, it is used in powderless etching.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-93-5
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00009260
InChI Key AFZZYIJIWUTJFO-UHFFFAOYSA-N
Synonym m-diethylbenzene, benzene, 1,3-diethyl, benzene, m-diethyl, m-ethylethylbenzene, unii-zm2x7i1g8y, zm2x7i1g8y, ethylated benzene, benzene, ethylated, meta-diethylbenzene, polyethylated benzene
PubChem CID 8864
IUPAC Name 1,3-diethylbenzene
SMILES CCC1=CC(=CC=C1)CC

Specifications

Density 0.86
Boiling Point 182°C
Flash Point 55°C (131°F)
Assay Percent Range >97%
Refractive Index 1.496
Quantity 10 mL
UN Number UN2049
Solubility Information Miscible with ethanol,benzene,carbon tetrachloride,ethyl ether and acetone. Immiscible with water and diethylbenzene.
Formula Weight 134.22
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 1, 3-Diethylbenzene
Show More Show Less
Hazard Category H226-H304-H315-H319-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIETHYLBENZENE
EINECSNumber 205-511-4
RTECSNumber CZ5620000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.