Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Difluorobenzene, 99+%, Thermo Scientific™

Click to view available options
Quantity:
10mL
25mL
100mL
Packaging:
Glass bottle
372-18-9
C6H4F2
114.10
MFCD00000327
UEMGWPRHOOEKTA-UHFFFAOYSA-N
m-difluorobenzene, benzene, 1,3-difluoro, benzene, m-difluoro, 2,4-difluorobenzene, meta-difluorobenzene, 1,3-difluoro benzene, unii-vo3c26woc2, 1,3-difluorbenzol, vo3c26woc2, 1,3difluorobenzene
9741
CHEBI:38584
1,3-difluorobenzene
FC1=CC(F)=CC=C1
372-18-9
C6H4F2
114.10
MFCD00000327
UEMGWPRHOOEKTA-UHFFFAOYSA-N
m-difluorobenzene, benzene, 1,3-difluoro, benzene, m-difluoro, 2,4-difluorobenzene, meta-difluorobenzene, 1,3-difluoro benzene, unii-vo3c26woc2, 1,3-difluorbenzol, vo3c26woc2, 1,3difluorobenzene
9741
CHEBI:38584
1,3-difluorobenzene
FC1=CC(F)=CC=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

CAS Min % 99.0
CAS Max % 100.0
Density 1.1500g/mL
Color Colorless to Yellow
Melting Point -59°C
Boiling Point 83°C
Flash Point 2°C
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula C6H4F2
Packaging Glass bottle
Beilstein 05, I, 108
Refractive Index 1.4370 to 1.439
Quantity 100mL
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,acetone,dmf,,2-propanol and methylene chloride
Specific Gravity 1.15
Formula Weight 114.09
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material 1, 3-Difluorobenzene
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-746-5

RUO – Research Use Only